accessibility menu, dialog, popup

UserName

CAS RN 7149-18-0

IUPAC Name: ethyl 2-aminocyclopent-1-ene-1-carboxylate
Synonyms: ethyl 2-aminocyclopent-1-ene-1-carboxylate
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: FMEHILPGLKGSCA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)CCC1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results