CAS RN 7126-39-8
IUPAC Name:
1H-pyrrole-3-carbaldehyde
Synonyms:
1H-pyrrole-3-carbaldehyde
Molecular Weight (g/mol):
95.1
Molecular Formula:
C5H5NO
InChi Key:
CHNYVNOFAWYUEG-UHFFFAOYSA-N
SMILES:
O=CC1=CNC=C1
1
–
1
of
1
results
1
Promotions Available
Pyrrole-3-carboxaldehyde, 97%
Pyrrole-3-carboxaldehyde, 97%
CAS: 7126-39-8 Molecular Formula: C5H5NO Molecular Weight (g/mol): 95.10 MDL Number: MFCD03426733 InChI Key: CHNYVNOFAWYUEG-UHFFFAOYSA-N Synonym: pyrrole-3-carboxaldehyde,1h-pyrrole-3-carboxaldehyde,pyrrole-3-carbaldehyde,3-formylpyrrole,3-formyl-1h-pyrrole,3-formyl-pyrrole,pyrrol-3-aldehyde,pyrrole-3-aldehyde,pubchem18051,acmc-209oht PubChem CID: 640308 IUPAC Name: 1H-pyrrole-3-carbaldehyde SMILES: O=CC1=CNC=C1