accessibility menu, dialog, popup

UserName

HOOO

CAS RN 710-11-2

IUPAC Name: 2-oxo-4-phenylbutanoic acid
Synonyms: 2-oxo-4-phenylbutanoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: PPKAIMDMNWBOKN-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)CCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results