accessibility menu, dialog, popup

UserName

CAS RN 710-04-3

IUPAC Name: 6-hexyloxan-2-one
Synonyms: delta-undecalactone
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: YZRXRLLRSPQHDK-UHFFFAOYNA-N
SMILES: CCCCCCC1CCCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results