accessibility menu, dialog, popup

UserName

O(S)OONH2N

CAS RN 70918-00-2

IUPAC Name: 4-[(2S)-2,3-dihydro-1,4-benzodioxine-2-carbonyl]piperazin-1-ium
Synonyms: 4-[(2S)-2,3-dihydro-1,4-benzodioxine-2-carbonyl]piperazin-1-ium
Molecular Weight (g/mol): 249.29
Molecular Formula: C13H17N2O3
InChi Key: FLUPDJNTYCSBJZ-LBPRGKRZSA-O
SMILES: O=C([C@@H]1COC2=CC=CC=C2O1)N1CC[NH2+]CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results