accessibility menu, dialog, popup

UserName

CAS RN 7085-85-0

IUPAC Name: ethyl 2-cyanoprop-2-enoate
Synonyms: ethyl cyanoacrylate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: FGBJXOREULPLGL-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=C)C#N
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results