accessibility menu, dialog, popup

UserName

CAS RN 70800-59-8

IUPAC Name: 2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Synonyms: 5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Molecular Weight (g/mol): 210.36
Molecular Formula: C4H2S5
InChi Key: NFKGILAHEQYFBG-UHFFFAOYSA-N
SMILES: S=C1SC2=C(SCS2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results