accessibility menu, dialog, popup

UserName

H3COOHOOH3C

CAS RN 708-76-9

IUPAC Name: 2-hydroxy-4,6-dimethoxybenzaldehyde
Synonyms: 2-hydroxy-4,6-dimethoxybenzaldehyde
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: FQRQWPNYJOFDLO-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C=O)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results