CAS RN 7064-38-2
IUPAC Name:
6-[(5Z)-5-({5-cyano-2-[3-(ethoxycarbonyl)piperidin-1-yl]-4-methyl-6-oxo-1-propyl-1,6-dihydropyridin-3-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]hexanoic acid
Synonyms:
6-[(5Z)-5-({5-cyano-2-[3-(ethoxycarbonyl)piperidin-1-yl]-4-methyl-6-oxo-1-propylpyridin-3-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]hexanoic acid
Molecular Weight (g/mol):
588.74
Molecular Formula:
C28H36N4O6S2
InChi Key:
GDARZPKSXSXRDC-JCMHNJIXNA-N
SMILES:
CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCCCCC(O)=O)C2=O)=C1N1CCCC(C1)C(=O)OCC
1
–
1
of
1
results
1
4-Iodo-5-methyl-isoxazole, 97%, Thermo Scientific™
CAS: 7064-38-2 Molecular Formula: C28H36N4O6S2 Molecular Weight (g/mol): 588.74 MDL Number: MFCD06411541 InChI Key: GDARZPKSXSXRDC-JCMHNJIXNA-N Synonym: 4-iodo-5-methylisoxazole,4-iodo-5-methyl-isoxazole,isoxazole,4-iodo-5-methyl,isoxazole, 4-iodo-5-methyl PubChem CID: 2795461 IUPAC Name: 4-iodo-5-methyl-1,2-oxazole SMILES: CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCCCCC(O)=O)C2=O)=C1N1CCCC(C1)C(=O)OCC