accessibility menu, dialog, popup

UserName

CAS RN 7064-33-7

CH3NOCH3NH3CCH3SSNOCH3OH3CON(Z)

CAS RN 7064-33-7

IUPAC Name: 5-{[(5Z)-3-[2-(3,4-dimethoxyphenyl)ethyl]-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-6-(3,5-dimethylpiperidin-1-yl)-4-methyl-2-oxo-1-propyl-1,2-dihydropyridine-3-carbonitrile
Synonyms: 5-{[(5Z)-3-[2-(3,4-dimethoxyphenyl)ethyl]-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-6-(3,5-dimethylpiperidin-1-yl)-4-methyl-2-oxo-1-propylpyridine-3-carbonitrile
Molecular Weight (g/mol): 594.79
Molecular Formula: C31H38N4O4S2
InChi Key: ALUPLTWVMIXEQH-DICXZTSXNA-N
SMILES: CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCC3=CC=C(OC)C(OC)=C3)C2=O)=C1N1CC(C)CC(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)