accessibility menu, dialog, popup

UserName

ClClClClClClClCl

CAS RN 706-78-5

IUPAC Name: octachlorocyclopent-1-ene
Synonyms: octachlorocyclopentene
Molecular Weight (g/mol): 343.66
Molecular Formula: C5Cl8
InChi Key: DMZRCHJVWAKCAX-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(Cl)(Cl)C(Cl)(Cl)C1(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results