CAS RN 7042-71-9
IUPAC Name:
4-(benzyloxy)-1H-indole-3-carbaldehyde
Synonyms:
4-(benzyloxy)-1H-indole-3-carbaldehyde
Molecular Weight (g/mol):
251.29
Molecular Formula:
C16H13NO2
InChi Key:
WURDDLFAUSUSLT-UHFFFAOYSA-N
SMILES:
O=CC1=CNC2=CC=CC(OCC3=CC=CC=C3)=C12
1
–
1
of
1
results
1
Promotions Available
4-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals
4-Benzyloxyindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals
CAS: 7042-71-9 Molecular Formula: C16H13NO2 Molecular Weight (g/mol): 251.29 MDL Number: MFCD00152002 InChI Key: WURDDLFAUSUSLT-UHFFFAOYSA-N Synonym: 4-benzyloxyindole-3-carboxaldehyde,4-benzyloxyindole-3-carbaldehyde,4-benzyloxy-1h-indole-3-carbaldehyde,4-phenylmethoxy-1h-indole-3-carboxaldehyde,4-phenylmethoxy indole-3-carbaldehyde,4-benayloxyindole-3-carboxaldehyde,pubchem7231,acmc-1bcm9,4-benzyloxy-3-formylindole,4-benzoxy-1h-indole-3-carbaldehyde PubChem CID: 7015846 IUPAC Name: 4-phenylmethoxy-1H-indole-3-carbaldehyde SMILES: O=CC1=CNC2=CC=CC(OCC3=CC=CC=C3)=C12