accessibility menu, dialog, popup

UserName

ONCl

CAS RN 70291-67-7

IUPAC Name: 4-(4-chlorophenyl)morpholine
Synonyms: 4-(4-chlorophenyl)morpholine
Molecular Weight (g/mol): 197.66
Molecular Formula: C10H12ClNO
InChi Key: KDWPYZPZUPWJGB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results