accessibility menu, dialog, popup

UserName

H3CPCH3H3CCH3CNNN

CAS RN 701921-71-3

IUPAC Name: [(cyanoimino)methylidene]azanide; trihexyl(tetradecyl)phosphanium
Synonyms: [(cyanoimino)methylidene]azanide; trihexyl(tetradecyl)phosphanium
Molecular Weight (g/mol): 549.91
Molecular Formula: C34H68N3P
InChi Key: DOMOOBQQQGXLGU-UHFFFAOYSA-N
SMILES: [N-]=C=NC#N.CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results