accessibility menu, dialog, popup

UserName

CAS RN 70170-23-9

CH3NOO

CAS RN 70170-23-9

IUPAC Name: 5-methyl-2-phenyl-1,3-oxazole-4-carbaldehyde
Synonyms: 5-methyl-2-phenyl-1,3-oxazole-4-carbaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: JEXONSMPSXTJFF-UHFFFAOYSA-N
SMILES: CC1=C(C=O)N=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)