accessibility menu, dialog, popup

UserName

CAS RN 7006-50-0

IUPAC Name: 1-benzyl-N-methylpiperidin-4-amine
Synonyms: 1-benzyl-N-methylpiperidin-4-amine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: RGEQSTMITLEXKD-UHFFFAOYSA-N
SMILES: CNC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results