accessibility menu, dialog, popup

UserName

CH3NOOHCH3(E/Z)

CAS RN 69891-38-9

IUPAC Name: N-(2,3-dimethylphenyl)-N-hydroxy-3-phenylprop-2-enamide
Synonyms: N-(2,3-dimethylphenyl)-N-hydroxy-3-phenylprop-2-enamide
Molecular Weight (g/mol): 267.33
Molecular Formula: C17H17NO2
InChi Key: SLIQYZYALKWWAT-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=CC=C1)N(O)C(=O)C=CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results