accessibility menu, dialog, popup

UserName

CAS RN 6980-08-1

IUPAC Name: 4-chloro-3-nitropyridin-2-amine
Synonyms: 4-chloro-3-nitropyridin-2-amine
Molecular Weight (g/mol): 173.56
Molecular Formula: C5H4ClN3O2
InChi Key: DIRINUVNYFAWQF-UHFFFAOYSA-N
SMILES: NC1=NC=CC(Cl)=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results