accessibility menu, dialog, popup

UserName

H3COOHOOH3C(E)

CAS RN 6972-61-8

IUPAC Name: (2E)-3-(2,4-dimethoxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2,4-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: YIKHDPHTFYWYJV-GQCTYLIASA-N
SMILES: COC1=CC=C(\C=C\C(O)=O)C(OC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results