CAS RN 6950-82-9
IUPAC Name:
2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetic acid
Synonyms:
(7-hydroxy-2-oxochromen-4-yl)acetic acid
Molecular Weight (g/mol):
220.18
Molecular Formula:
C11H8O5
InChi Key:
BNHPMQBVNXMPDU-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(=O)OC2=CC(O)=CC=C12
1
–
1
of
1
results
1
Promotions Available
7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
CAS: 6950-82-9 Molecular Formula: C11H8O5 Molecular Weight (g/mol): 220.18 MDL Number: MFCD00037563 InChI Key: BNHPMQBVNXMPDU-UHFFFAOYSA-N Synonym: 7-hydroxycoumarin-4-acetic acid,2-7-hydroxy-2-oxo-2h-chromen-4-yl acetic acid,umbelliferone-4-acetic acid,7-hydroxy-2-oxo-2h-1-benzopyran-4-acetic acid,chembl12253,2-7-hydroxy-2-oxochromen-4-yl acetic acid,7-hydroxy-4-coumarinylacetic acid,2h-1-benzopyran-4-acetic acid, 7-hydroxy-2-oxo,7-hydroxycoumarin-4-yl acetic acid,7-hydroxy-2-oxochromen-4-yl acetic acid PubChem CID: 5338490 IUPAC Name: 2-(7-hydroxy-2-oxochromen-4-yl)acetic acid SMILES: C1=CC2=C(C=C1O)OC(=O)C=C2CC(=O)O