accessibility menu, dialog, popup

UserName

CAS RN 69491-56-1

NH2NN

CAS RN 69491-56-1

IUPAC Name: 3-(pyrimidin-2-yl)aniline
Synonyms: 3-(pyrimidin-2-yl)aniline
Molecular Weight (g/mol): 171.2
Molecular Formula: C10H9N3
InChi Key: GOCBKTKLGFLECW-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)