CAS RN 69464-98-8
IUPAC Name:
4-bromo-3-methyl-1-phenyl-1H-pyrazol-5-amine
Synonyms:
4-bromo-5-methyl-2-phenylpyrazol-3-amine
Molecular Weight (g/mol):
252.12
Molecular Formula:
C10H10BrN3
InChi Key:
KNUGDEKPCNCMIO-UHFFFAOYSA-N
SMILES:
CC1=NN(C(N)=C1Br)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Amino-4-bromo-3-methyl-1-phenylpyrazole 96.0+%, TCI America™
CAS: 69464-98-8 Molecular Formula: C10H10BrN3 Molecular Weight (g/mol): 252.115 MDL Number: MFCD04039926 InChI Key: KNUGDEKPCNCMIO-UHFFFAOYSA-N Synonym: 4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-amine,4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-ylamine,5-amino-4-bromo-3-methyl-1-phenylpyrazole,acmc-20amn6,4-bromo-5-methyl-2-phenyl-2h-pyrazol-3,5-amino-4-bromo-3-methyl-1-phenyl-1h-pyrazole,4-bromo-5-methyl-2-phenyl-2h-pyrazol-3-ylamine,4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-,1h-pyrazol-5-amine, 4-bromo-3-methyl-1-phenyl PubChem CID: 4339679 IUPAC Name: 4-bromo-5-methyl-2-phenylpyrazol-3-amine SMILES: CC1=NN(C(=C1Br)N)C2=CC=CC=C2