accessibility menu, dialog, popup

UserName

H3COH3CN

CAS RN 6940-57-4

IUPAC Name: 1-(6-methylpyridin-2-yl)ethan-1-one
Synonyms: 1-(6-methylpyridin-2-yl)ethanone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: FPQMUQPPAYCAME-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results