accessibility menu, dialog, popup

UserName

CH3SN

CAS RN 693-95-8

IUPAC Name: 4-methyl-1,3-thiazole
Synonyms: 4-methylthiazole
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: QMHIMXFNBOYPND-UHFFFAOYSA-N
SMILES: CC1=CSC=N1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)