accessibility menu, dialog, popup

UserName

CAS RN 6913-23-1

OSSHN

CAS RN 6913-23-1

IUPAC Name: 2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: rhodanine
Molecular Weight (g/mol): 133.18
Molecular Formula: C3H3NOS2
InChi Key: KIWUVOGUEXMXSV-UHFFFAOYSA-N
SMILES: O=C1CSC(=S)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results