accessibility menu, dialog, popup

UserName

NiClCl

CAS RN 69098-15-3

IUPAC Name: nickel(2+) dichloride
Synonyms: nickel chloride
Molecular Weight (g/mol): 129.59
Molecular Formula: Cl2Ni
InChi Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ni++]
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)