accessibility menu, dialog, popup

UserName

CAS RN 68844-77-9

IUPAC Name: 1-[(4-fluorophenyl)methyl]-N-{1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl}-1H-1,3-benzodiazol-2-amine
Synonyms: astemizole janssen brand of astemizole astemizole esteve brand fustery brand of astemizole lesvi brand of astemizole rifedot urdrim astemizole ratiopharm brand astemizole lesvi brand diba brand of astemizole
Molecular Weight (g/mol): 458.58
Molecular Formula: C28H31FN4O
InChi Key: GXDALQBWZGODGZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCN2CCC(CC2)NC2=NC3=CC=CC=C3N2CC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results