accessibility menu, dialog, popup

UserName

CAS RN 68553-60-6

OOOOOOOOV

CAS RN 68553-60-6

IUPAC Name: vanadium(4+) tetrakis(naphthalene-2-carboxylate)
Synonyms: vanadium(4+) tetrakis(2-naphthoate)
Molecular Weight (g/mol): 735.64
Molecular Formula: C44H28O8V
InChi Key: DLHFCHPXIYNOOK-UHFFFAOYSA-J
SMILES: [V+4].[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results