accessibility menu, dialog, popup

UserName

CAS RN 685508-28-5

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}cyclobutane-1-carboxylic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}cyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 249.27
Molecular Formula: C13H15NO4
InChi Key: ZNRUEEQIUVKKBL-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC1NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results