accessibility menu, dialog, popup

UserName

CH3NOONHO

CAS RN 68550-75-4

IUPAC Name: N-cyclohexyl-N-methyl-4-[(2-oxo-1,2-dihydroquinolin-6-yl)oxy]butanamide
Synonyms: cilostamide
Molecular Weight (g/mol): 342.44
Molecular Formula: C20H26N2O3
InChi Key: UIAYVIIHMORPSJ-UHFFFAOYSA-N
SMILES: CN(C1CCCCC1)C(=O)CCCOC1=CC=C2NC(=O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results