accessibility menu, dialog, popup

UserName

CAS RN 6841-96-9

IUPAC Name: 2-methyl-2-nitrosopropane
Synonyms: 2-methyl-2-nitrosopropane
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: LUQZKEZPFQRRRK-UHFFFAOYSA-N
SMILES: CC(C)(C)N=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results