accessibility menu, dialog, popup

UserName

H3CCH3H3CN

CAS RN 68141-13-9

IUPAC Name: 6-tert-butylquinoline
Synonyms: 6-tert-butylquinoline
Molecular Weight (g/mol): 185.27
Molecular Formula: C13H15N
InChi Key: JHAWWJQGHKGXHA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C2N=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results