accessibility menu, dialog, popup

UserName

OSSSS

CAS RN 68128-94-9

IUPAC Name: 2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole-4-carbaldehyde
Synonyms: 2-(1,3-dithiol-2-ylidene)-1,3-dithiole-4-carbaldehyde
Molecular Weight (g/mol): 232.35
Molecular Formula: C7H4OS4
InChi Key: NDJHQJARYXJFDI-UHFFFAOYSA-N
SMILES: O=CC1=CSC(S1)=C1SC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results