accessibility menu, dialog, popup

UserName

CAS RN 68105-93-1

ClBr

CAS RN 68105-93-1

IUPAC Name: 1-bromo-7-chloroheptane
Synonyms: 1-bromo-7-chloroheptane
Molecular Weight (g/mol): 213.54
Molecular Formula: C7H14BrCl
InChi Key: ZMAOWMBSLZYILL-UHFFFAOYSA-N
SMILES: ClCCCCCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results