CAS RN 680-65-9
IUPAC Name:
1,3-diethyl 2,2-difluoropropanedioate
Synonyms:
1,3-diethyl 2,2-difluoropropanedioate
Molecular Weight (g/mol):
196.15
Molecular Formula:
C7H10F2O4
InChi Key:
WCRVWLHTROHXBB-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(F)(F)C(=O)OCC
1
–
1
of
1
results
1
Promotions Available
Diethyl difluoromalonate, 97%
Diethyl difluoromalonate, 97%
CAS: 680-65-9 Molecular Formula: C7H10F2O4 Molecular Weight (g/mol): 196.15 MDL Number: MFCD00221617 InChI Key: WCRVWLHTROHXBB-UHFFFAOYSA-N Synonym: diethyl 2,2-difluoromalonate,diethyl difluoromalonate,2,2-difluoro-malonic acid diethyl ester,1,3-diethyl 2,2-difluoropropanedioate,difluoromalonic acid diethyl ester,diethyl2,2-difluoromalonate,propanedioic acid, 2,2-difluoro-, 1,3-diethyl ester,propanedioic acid, difluoro-, diethyl ester,diethyl 2,2-difluoromalonate #,diethyl2,2-difluoropropanedioatev PubChem CID: 260648 IUPAC Name: diethyl 2,2-difluoropropanedioate SMILES: CCOC(=O)C(F)(F)C(=O)OCC