CAS RN 680-05-7
CAS RN 680-05-7
IUPAC Name:
methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate
Synonyms:
methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate
Molecular Weight (g/mol):
228.07
Molecular Formula:
C5H3F7O2
InChi Key:
CGMUKBZUGMXXEF-UHFFFAOYSA-N
SMILES:
COC(=O)C(F)(C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Methyl Heptafluoroisobutyrate 98.0+%, TCI America™
CAS: 680-05-7 Molecular Formula: C5H3F7O2 Molecular Weight (g/mol): 228.066 MDL Number: MFCD00633403 InChI Key: CGMUKBZUGMXXEF-UHFFFAOYSA-N Synonym: Heptafluoroisobutyric Acid Methyl Ester PubChem CID: 2775518 IUPAC Name: methyl 2,3,3,3-tetrafluoro-2-(trifluoromethyl)propanoate SMILES: COC(=O)C(C(F)(F)F)(C(F)(F)F)F