accessibility menu, dialog, popup

UserName

CAS RN 6790-09-6

HOOOOOOOOOOOOOH

CAS RN 6790-09-6

IUPAC Name: 3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane-1,35-diol
Synonyms: dodecaethylene glycol
Molecular Weight (g/mol): 546.65
Molecular Formula: C24H50O13
InChi Key: WRZXKWFJEFFURH-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results