accessibility menu, dialog, popup

UserName

H3COOH3CCH3(E)(Z)

CAS RN 67883-79-8

IUPAC Name: (3E)-hex-3-en-1-yl (2Z)-2-methylbut-2-enoate
Synonyms: cis-3-hexenyl tiglate
Molecular Weight (g/mol): 182.26
Molecular Formula: C11H18O2
InChi Key: JNWQKXUWZWKUAY-BAGHCZIJSA-N
SMILES: CC\C=C\CCOC(=O)C(\C)=C/C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results