CAS RN 67883-62-9
IUPAC Name:
tributyl(2-methylprop-2-en-1-yl)stannane
Synonyms:
tributyl(2-methylprop-2-en-1-yl)stannane
Molecular Weight (g/mol):
345.16
Molecular Formula:
C16H34Sn
InChi Key:
CYIUFVYZGOCGEV-UHFFFAOYSA-N
SMILES:
CCCC[Sn](CCCC)(CCCC)CC(C)=C
1
–
1
of
1
results
1
Promotions Available
Methallyltri-n-butyltin, 98%
Methallyltri-n-butyltin, 98%
CAS: 67883-62-9 Molecular Formula: C16H34Sn Molecular Weight (g/mol): 345.158 MDL Number: MFCD03425867 InChI Key: CYIUFVYZGOCGEV-UHFFFAOYSA-N Synonym: methallyltri-n-butyltin,tributyl 2-methylallyl stannane,stannane, tributyl 2-methyl-2-propenyl,tributyl 2-methylprop-2-en-1-yl stannane,methallyltributyltin,2-methylallyltributyltin,2-methylallyl tributyltin,methallyltri-n-butylstannane,methallyltri-n-butylin,2-methylallyl-tributylstannane PubChem CID: 11013379 IUPAC Name: tributyl(2-methylprop-2-enyl)stannane SMILES: CCCC[Sn](CCCC)(CCCC)CC(=C)C