accessibility menu, dialog, popup

UserName

CAS RN 676525-77-2

CH3H3CCH3NNCH3H3CCH3CNCNFPFFFFFIr

CAS RN 676525-77-2

IUPAC Name: iridium(3+) bis(2-(pyridin-2-yl)benzen-1-ide) 4,4'-di-tert-butyl-2,2'-bipyridine hexafluoro-λ⁵-phosphanuide
Synonyms: iridium(3+) bis(2-(pyridin-2-yl)benzen-1-ide) 4,4'-di-tert-butyl-2,2'-bipyridine hexafluorophosphate
Molecular Weight (g/mol): 913.97
Molecular Formula: C40H40F6IrN4P
InChi Key: VCIVELSSYHAWGC-UHFFFAOYSA-N
SMILES: [Ir+3].F[P-](F)(F)(F)(F)F.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1.CC(C)(C)C1=CC(=NC=C1)C1=NC=CC(=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results