accessibility menu, dialog, popup

UserName

NFFOHOF

CAS RN 675602-91-2

IUPAC Name: 2,3,5-trifluoropyridine-4-carboxylic acid
Synonyms: 2,3,5-trifluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 177.08
Molecular Formula: C6H2F3NO2
InChi Key: UKLNELOJJKZMLJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=NC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results