accessibility menu, dialog, popup

UserName

CAS RN 67526-95-8

IUPAC Name: (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
Synonyms: thapsigargin thapsigargine
Molecular Weight (g/mol): 650.76
Molecular Formula: C34H50O12
InChi Key: IXFPJGBNCFXKPI-FSIHEZPISA-N
SMILES: CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC
Physical Form 
  • (1)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)