accessibility menu, dialog, popup

UserName

CAS RN 674773-07-0

SNNOOOOOOOOH2OSrSr

CAS RN 674773-07-0

IUPAC Name: distrontium(2+) 5-[bis(carboxylatomethyl)amino]-3-(carboxylatomethyl)-4-cyanothiophene-2-carboxylate hydrate
Synonyms: distrontium(2+) 5-[bis(carboxylatomethyl)amino]-3-(carboxylatomethyl)-4-cyanothiophene-2-carboxylate hydrate
Molecular Weight (g/mol): 531.5
Molecular Formula: C12H8N2O9SSr2
InChi Key: KXBBXBBUNMMNSU-UHFFFAOYSA-J
SMILES: O.[Sr++].[Sr++].[O-]C(=O)CN(CC([O-])=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results