CAS RN 669773-34-6
CAS RN 669773-34-6
IUPAC Name:
6,10-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms:
6,10-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol):
342.05
Molecular Formula:
C12H6Br2S
InChi Key:
ULGFJZPCGNTWFK-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC2=C1SC1=C2C=CC=C1Br
1
–
1
of
1
results
1
4,6-Dibromodibenzothiophene 98.0+%, TCI America™
CAS: 669773-34-6 Molecular Formula: C12H6Br2S Molecular Weight (g/mol): 342.05 MDL Number: MFCD18451473 InChI Key: ULGFJZPCGNTWFK-UHFFFAOYSA-N Synonym: 4,6-dibromodibenzo b,d thiophene,4,6-dibromo-dibenzothiophene,6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene,dibenzothiophene, 4,6-dibromo,6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene PubChem CID: 5245996 IUPAC Name: 6,10-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene SMILES: BrC1=CC=CC2=C1SC1=C2C=CC=C1Br