accessibility menu, dialog, popup

UserName

H3CO

CAS RN 6697-12-7

IUPAC Name: 1-phenylhexadecan-1-one
Synonyms: hexadecanophenone
Molecular Weight (g/mol): 316.53
Molecular Formula: C22H36O
InChi Key: IIOLAWJMOGLOIB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results