accessibility menu, dialog, popup

UserName

OHNSNH2

CAS RN 66892-25-9

IUPAC Name: [(oxolan-2-yl)methyl]thiourea
Synonyms: oxolan-2-ylmethylthiourea
Molecular Weight (g/mol): 160.24
Molecular Formula: C6H12N2OS
InChi Key: DYYSJNQVLWANCC-UHFFFAOYNA-N
SMILES: NC(=S)NCC1CCCO1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results