accessibility menu, dialog, popup

UserName

CAS RN 66492-51-1

IUPAC Name: (5-ethyl-1,3-dioxan-5-yl)methyl prop-2-enoate
Synonyms: (5-ethyl-1,3-dioxan-5-yl)methyl prop-2-enoate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: STGXUBIZGYMIRM-UHFFFAOYSA-N
SMILES: CCC1(COC(=O)C=C)COCOC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results