accessibility menu, dialog, popup

UserName

CAS RN 66137-74-4

FFIFFOFFFFSFOO

CAS RN 66137-74-4

IUPAC Name: 1,1,2,2-tetrafluoro-2-(1,1,2,2-tetrafluoro-2-iodoethoxy)ethane-1-sulfonyl fluoride
Synonyms: 1,1,2,2-tetrafluoro-2-(1,1,2,2-tetrafluoro-2-iodoethoxy)ethanesulfonyl fluoride
Molecular Weight (g/mol): 425.99
Molecular Formula: C4F9IO3S
InChi Key: XSLYISNQTJHKMP-UHFFFAOYSA-N
SMILES: FC(F)(I)C(F)(F)OC(F)(F)C(F)(F)S(F)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results