accessibility menu, dialog, popup

UserName

CAS RN 66-23-9

IUPAC Name: [2-(acetyloxy)ethyl]trimethylazanium bromide
Synonyms: acetylcholine bromide tonocholin B bromoacetylcholine acetylcholine hydrobromide pragmoline acetylcholine bromhydrate
Molecular Weight (g/mol): 226.11
Molecular Formula: C7H16BrNO2
InChi Key: ZEHGKSPCAMLJDC-UHFFFAOYSA-M
SMILES: [Br-].CC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results