accessibility menu, dialog, popup

UserName

CAS RN 65548-55-2

IUPAC Name: 2-(3,4-dimethoxyphenyl)-7,8-dimethoxy-4H-chromen-4-one
Synonyms: 2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
Molecular Weight (g/mol): 342.35
Molecular Formula: C19H18O6
InChi Key: ZRLWYUNKZNRQLO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C1=CC(=O)C2=CC=C(OC)C(OC)=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results